missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver 2,4-pentanedionate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Silver 2,4-pentanedionate is used to make silver impregnated nanocrystalline metal oxides, with special emphasis on aluminum oxide. It is used as a cationization reagent in combination with polar matrices such as 2,5-dihydroxybenzoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Silver 2,4-pentanedionate is used to make silver impregnated nanocrystalline metal oxides, with special emphasis on aluminum oxide. It is used as a cationization reagent in combination with polar matrices such as 2,5-dihydroxybenzoic acid.
Solubility
Soluble in water.
Notes
Air, light and moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
Silver 2,4-pentanedionate is used to make silver impregnated nanocrystalline metal oxides, with special emphasis on aluminum oxide. It is used as a cationization reagent in combination with polar matrices such as 2,5-dihydroxybenzoic acid.
Solubility
Soluble in water.
Notes
Air, light and moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 100°C (decomposition) |
Quantity | 1 g |
Solubility Information | Soluble in water. |
Formula Weight | 206.98 |
Percent Purity | 98% |
Odor | Characteristic |
Sensitivity | Air, light, and moisture sensitive |
Chemical Name or Material | Silver 2,4-pentanedionate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'