Learn More
Trajan Scientific and Medical™ SilTite™ Metal Nuts
SilTite FingerTite delivers an easy, leak-free installation for capillary columns without the use of any tools
Brand: Trajan Scientific and Medical™ 073232
Code : 13
Additional Details : Weight : 0.10000kg
Description
- Next generation ferrule system for gas chromatography
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifications
Shimadzu GC14 A, GC17 A, GC2010 and GC2014 Detector/Injectors | |
SilTite | |
Nut | |
5/Pk. |