missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Silicone Stoppers
Silicone construction with natural color. Prevents spillage while working in the lab.
Brand: BRAND™ 144330
Code : NEW
Additional Details : Weight : 0.82000kg
Description
- Silicone rubber construction
- Used to prevent inadvertant leaks and spills
- Natural color
Specifications
Natural | |
17 mm | |
Silicone |
25 mm | |
22 mm |