missing translation for 'onlineSavingsMsg'
Learn More

Silicon sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 15404825
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

 

Silicon sputtering target in ultra high purity silicon is used in the semiconductor industry as a result of its semiconducting properties. It is used as an alloying element in the manufacture of certain alloys (e.g. ferrosilicon, an alloy of iron and silicon which is used to introduce silicon into steel and cast iron). It is also used in the manufacture of glass and computer microchips.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 Ea.
CAS 7440-21-3
Odor Odorless
Molecular Formula Si
MDL Number MFCD00085311
Synonym metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
Solubility Information Insoluble in water.
InChI Key XUIMIQQOPSSXEZ-UHFFFAOYSA-N
SMILES [Si]
IUPAC Name silicon
Molecular Weight (g/mol) 28.09
PubChem CID 5461123
ChEBI CHEBI:27573
Percent Purity 99.999%
Assay (metals basis)
Chemical Name or Material Silicon sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'