missing translation for 'onlineSavingsMsg'
Learn More

Silicon rod, 5.08cm (2.0 in.) dia., 99.999% (metals basis), Thermo Scientific™

Catalog Number p-7050614
Click to view available options
Quantity:
25 mm
100 mm
This item is not returnable. View return policy
7440-21-3
Si
28.09
MFCD00085311
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
5461123
CHEBI:27573
silicon
[Si]
This item is not returnable. View return policy
7440-21-3
Si
28.09
MFCD00085311
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
5461123
CHEBI:27573
silicon
[Si]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 25 mm
Percent Purity 99.999%
Odor Odorless
Weight ≈47.18g/cm
Form Rod
Assay (metals basis)
Chemical Name or Material Silicon rod, 5.08cm (2.0 in.) dia.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'