missing translation for 'onlineSavingsMsg'
Learn More

Silicon powder, crystalline, -140 mesh, 98% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7050907
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
7440-21-3
Si
28.09
MFCD00085311
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
5461123
CHEBI:27573
silicon
[Si]
This item is not returnable. View return policy
7440-21-3
Si
28.09
MFCD00085311
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
5461123
CHEBI:27573
silicon
[Si]

For plasma spraying

For plasma spraying

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray
UN Number UN1346
Quantity 50 g
Percent Purity 98%
Mesh Size −140 Mesh
Odor Odorless
Form Crystalline Powder
Assay (metals basis)
Chemical Name or Material Silicon powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'