missing translation for 'onlineSavingsMsg'
Learn More

Silicon powder, crystalline, -100+325 mesh, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7050222
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
7440-21-3
Si
28.09
MFCD00085311
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
5461123
CHEBI:27573
silicon
[Si]
This item is not returnable. View return policy
7440-21-3
Si
28.09
MFCD00085311
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
5461123
CHEBI:27573
silicon
[Si]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
UN Number UN1346
Quantity 10 g
Percent Purity 99.999%
Mesh Size −100+325 Mesh
Odor Odorless
Form Crystalline Powder
Assay (metals basis)
Chemical Name or Material Silicon powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'