missing translation for 'onlineSavingsMsg'
Learn More

Silicon, plasma standard solution, Specpure™ Si 10,000μg/mL, Thermo Scientific Chemicals

Catalog Number p-7050582
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy
MFCD00085311
This item is not returnable. View return policy
MFCD00085311

Silicon plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 50 mL
Grade Specpure™
Solubility Miscible with molten alkali oxides, a mixture of nitric and hydrofluoric acids; Immiscible with water, nitric and hydrochloric acid
Physical Form Liquid
Chemical Name or Material Silicon plasma standard solution, Si 10000μg/mL

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'