missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon(IV) oxide sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.995% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 Ea.
Description
Magnetron sputtering source
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 1 Ea. |
MDL Number | MFCD00011232 |
Formula Weight | 60.09 |
Percent Purity | 99.995% |
Assay | (metals basis) |
Chemical Name or Material | Silicon(IV) oxide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'