missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon(IV) oxide, 99.5% (metals basis), Thermo Scientific Chemicals
50.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
Description
Silicon(IV) oxide is used in the manufacture of glass for windows, drinking glasses, beverage bottles and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 2230°C |
Melting Point | 1710°C |
Color | Brown to White |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 99.5% (metals basis) |
Solubility Information | Insoluble in water and acids. |
IUPAC Name | silanedione |
Formula Weight | 60.09 |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'