missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) oxide, microcrystalline, Thermo Scientific Chemicals

Catalog Number p-4407785
36.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
2 kg
10 kg
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
14808-60-7
O2Si
60.08
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
24261
CHEBI:30563
O=[Si]=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
14808-60-7
O2Si
60.08
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
24261
CHEBI:30563
O=[Si]=O

In the manufacture of glass, ceramics and abrasives, in water filtration, in cosmetics, pharmaceuticals and rubber, as a thermal insulatorSilica is used in the manufacture of glass for windows, drinking glasses, beverage bottles, and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware, and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 2230°C
Melting Point 1710°C
Color Brown to White
Physical Form Powder
Quantity 2 kg
Assay Percent Range 99.5%
Solubility Information Insoluble in water and acids. Soluble in HF. Very slightly soluble in alkalies
IUPAC Name dioxosilane
Formula Weight 60.09
Odor Odorless
Chemical Name or Material Silicon(IV) oxide, 98.5-99% microcrystalline silicon in the form of quartz
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'