missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon(IV) 2-ethylhexanoate, nominally 75% in ethanol, Thermo Scientific Chemicals
362.68 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
Description
In the manufacture of its esters for artificial flavors, and in the manufacture of hexyl derivatives, such as hexylphenols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS | 67939-81-5 |
Color | Yellow |
Molecular Formula | C32H60O8Si |
MDL Number | MFCD00269842 |
Quantity | 250 g |
UN Number | UN1170 |
Sensitivity | Moisture sensitive |
Synonym | silicon 2-ethylhexanoate, tetrakis 2-ethylhexanoic acid, tetraanhydride with silicic acid, 2-ethylhexanoic acid tetraanhydride with silicic acid h4sio4, hexanoic acid, 2-ethyl-, 1,1',1,1'-tetraanhydride with silicic acid h4sio4, hexanoic acid, 2-ethyl-, tetraanhydride with silicic acid h4sio4, silicon 2-ethylhexanoate, min. 90, tris 2-ethylhexanoyloxy silyl 2-ethylhexanoate, silicon iv 2-ethylhexanoate in ethanol, silicic acid tetrakis 2-ethylhexanoic tetraanhydride, silicon iv 2-ethylhexanoate, nominally in ethanol |
Solubility Information | Hydrolyzes in water. |
InChI Key | UNKOLEUDCQIVBV-UHFFFAOYSA-N |
Show More |
For Manufacturing and Laboratory Use. Not for beverage use
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'