missing translation for 'onlineSavingsMsg'
Learn More

Silicon(II) oxide, Optical Grade, 99.7% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7080489
Click to view available options
Quantity:
250 g
This item is not returnable. View return policy
10097-28-6
OSi
44.084
MFCD00151536
LIVNPJMFVYWSIS-UHFFFAOYSA-N
silicon monoxide, silylene, oxo, silicon ii oxide, unii-1oqn9cbg7l, 1oqn9cbg7l, silicon ii oxide, optical grade, sio, oxidosilicon, oxosilanylidene, oxosilylene
66241
CHEBI:30588
oxoniumylidynesilanide
[O+]#[Si-]
This item is not returnable. View return policy
10097-28-6
OSi
44.084
MFCD00151536
LIVNPJMFVYWSIS-UHFFFAOYSA-N
silicon monoxide, silylene, oxo, silicon ii oxide, unii-1oqn9cbg7l, 1oqn9cbg7l, silicon ii oxide, optical grade, sio, oxidosilicon, oxosilanylidene, oxosilylene
66241
CHEBI:30588
oxoniumylidynesilanide
[O+]#[Si-]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1880°C
Melting Point >1702°C
Color Black
Quantity 250 g
Linear Formula SiO
Merck Index 14,8496
Formula Weight 44.09
Percent Purity 99.7%
Grade Optical
Odor Odorless
Density 2.13 g/mL
Assay (metals basis)
Chemical Name or Material Silicon (II) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'