missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Silicon(II) oxide, 99.9% (metals basis)

For vacuum deposition

Brand:  Alfa Aesar™ 041711.36

Code : NEW

Additional Details : CAS Number : 10097-28-6 Weight : 0.50000kg

Quantity 500g

 View more versions of this product

Product Code. 11318858

  • £424.90 / 500g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

Silicon(II) oxide
6 to 15mm Pieces
MFCD00151536
Oxosilylene
10097-28-6
44.09
14,8496

Specifications

Silicon(II) oxide
10097-28-6
99.9%
44.09
14,8496
2.13g/mL
1880°C
500g
6 to 15mm Pieces
Pieces
(metals basis)
MFCD00151536
Oxosilylene
Odorless
>1702°C
Videos
SDS