missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Chiralplate™ for TLC Enantiomer Separation
Resolve a wide variety of enantiomers. Chiralplate™ for TLC Enantiomer Separation consists of a special layer available on glass plates with fluorescent indicator.
Brand: Macherey-Nagel™ 811059
Code : 50
Additional Details : Weight : 2.00000kg
Description
Features- Reversed phase nano silica impregnated with Cu(II) ions and a chiral selector (a proline derivative, DP 31 43 726 and EP 0 143 147)
- Separation based on ligand exchange, i. e. formation of ternary mixed-ligand complexes with the Cu(II) ions; differences in the stability of the diastereomeric complexes cause chromatographic separation
- Available as glass plates with fluorescent indicator
Application
- Enantiomer separation of amino acids, N-methylamino acids, N-formylamino acids, α-alkylamino acids, thiazolidine derivatives, dipeptides, lactones, α-hydroxycarboxylic acids
- A review on the application of CHIRALPLATE has been given by K. Günther [J. Chromatogr. 448 (1988) 11–30].
Specifications
Silica | |
4 | |
UV254 | |
0.25 mm |
Silicon | |
Glass Plate | |
For TLC Enantiomer Separation | |
10 x 20 cm |