missing translation for 'onlineSavingsMsg'
Learn More

SGE™ Silica Tubing

Brand:  SGE™ 064033

Code : NEW

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15248853

  • £197.00 / 10m


Estimated Shipment
22-02-2018
Add to basket

Specifications

Specifications

0.68mm
Silica
10m
Tubing
Videos