missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan™ Methyl Deactivated Fused Silica Capillary Tubing
Essential for maintaining a high performance chromatography system.Trajan Scientific and Medical™ Methyl Deactivated Fused Silica Capillary Tubing is chemically inert and thermally stable up to 380°C.
Brand: Trajan™ 0624431
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Suitable for organic and aqueous solvents
- Methyl deactivated for use with hydrocarbons (pentane, hexane, heptane, iso-octane, aromatics and mixed solvents - n-paraffins/chlorinated solvents
- Individually tested performance of each 30m length from which all shorter lengths are prepared
- Ideal for biotechnology applications
Specifications
Capillary Tubing | |
0.01 in. | |
0.363 mm | |
2 m |
0.25 mm | |
0.001 in. | |
6.6 ft. | |
Silica |