Learn More
Thermo Scientific Chemicals Silica gel, wide pore, 150 angstroms
£28.20 - £1251.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11375497
|
Thermo Scientific Chemicals
042726.18 |
50 g |
£28.20
50g |
In Stock
Log in to see stock. |
|||||
11385497
|
Thermo Scientific Chemicals
042726.30 |
250 g |
£106.00
250g |
In Stock
Log in to see stock. |
|||||
11395497
|
Thermo Scientific Chemicals
042726.A1 |
1 kg |
£316.00
1kg |
In Stock
Log in to see stock. |
|||||
11365497
|
Thermo Scientific Chemicals
042726.A7 |
5 kg |
£1251.00
5kg |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
Description
Silica gel, wide pore is mainly used for the dehydrated purification of the industrial gases, the clearance of the organic acids and high polymers in the insulative oil, the purification and separation of the medicines. It is also used in column chromatography without applied pressure (gravity pulled).