Learn More
Alfa Aesar™ Silica gel, preparative chromatography grade, spherical, 15 micron APS, 60 angstroms, 99.99+%
Brand:  Alfa Aesar™ 045824.06
Code : NEW
Additional Details : CAS Number : 63231-67-4 Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15404985
  • £25.76 / 5g
Estimated Shipment date
from Supplier 06-02-2018
Add to basket
Specifications
Chemical Identifiers
Silica gel
P.V. 1.00cc/g
MFCD00011232
63231-67-4
Preparative Chromatography Grade
Specifications
Silica gel
P.V. 1.00cc/g
Preparative Chromatography Grade
Spherical
MFCD00011232
5g
63231-67-4
15 Micron APS, 60Å
≥99.99%
650m2/g