Learn More
Alfa Aesar™ Silica gel, HPLC grade, spherical, 5 micron APS, 260 angstroms, 99.99+%
Brand:  Alfa Aesar™ 045812.06
Code : NEW
Additional Details : CAS Number : 63231-67-4 Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15417095
  • £78.02 / 5g
Estimated Shipment date
from Supplier 25-05-2018
Add to basket
Specifications
Chemical Identifiers
Silica gel
P.V. 0.90cc/g
MFCD00011232
63231-67-4
HPLC Grade
Specifications
Silica gel
P.V. 0.90cc/g
HPLC Grade
Spherical
MFCD00011232
5g
63231-67-4
3 Micron APS, 260Å
≥99.99%
150m2/g