missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, HPLC grade, spherical, 3 micron APS, 120 angstroms, 99.99+%, Thermo Scientific Chemicals

Catalog Number p-7059958
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
63231-67-4
H2O3Si
78.098
MFCD00011232
IJKVHSBPTUYDLN-UHFFFAOYSA-N
14768
dihydroxy(oxo)silane
O[Si](=O)O
This item is not returnable. View return policy
63231-67-4
H2O3Si
78.098
MFCD00011232
IJKVHSBPTUYDLN-UHFFFAOYSA-N
14768
dihydroxy(oxo)silane
O[Si](=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Spherical, S.A. 340m
Material Silica Gel
Quantity 1 g
Particle Size 3 Micron APS, 120Å
Percent Purity ≥99.99%
Grade HPLC
Surface Area 340m2/g
Chemical Name or Material Silica gel, P.V. 1.00cc/g

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'