Learn More
Alfa Aesar™ Silica gel, HPLC grade, spherical, 3 micron APS, 120 angstroms, 99.99+%
Brand:  Alfa Aesar™ 045803.14
Code : NEW
Additional Details : CAS Number : 63231-67-4 Weight : 0.02500kg
Quantity 25g
 View more versions of this product
Product Code. 15437085
  • £584.10 / 25g
Estimated Shipment date
from Supplier 07-03-2018
Add to basket
Specifications
Chemical Identifiers
Silica gel
P.V. 1.00cc/g
MFCD00011232
63231-67-4
HPLC Grade
Specifications
Silica gel
P.V. 1.00cc/g
HPLC Grade
Spherical
MFCD00011232
25g
63231-67-4
3 Micron APS, 120Å
≥99.99%
340m2/g