missing translation for 'onlineSavingsMsg'
Learn More

Silica gel 60, 0.019-0.037mm (400-600 mesh), Thermo Scientific Chemicals

Catalog Number p-4408665
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
This item is not returnable. View return policy
7631-86-9
O2Si
60.08
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
CHEBI:30563
silanedione
O=[Si]=O
This item is not returnable. View return policy
7631-86-9
O2Si
60.08
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
CHEBI:30563
silanedione
O=[Si]=O

AA4272918

Column chromatography with elevated pressureSilica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Color White
Quantity 50 g
Solubility Information Soluble in hydrofluoric acid. Insoluble in water.
Odor Odorless
Chemical Name or Material Silica gel 60

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'