missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica Gel, 60 to 200 Mesh, BAKER ANALYZED™ Reagent, J.T. Baker™
555.75 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
Description
- Suitable For Column Chromatography
Specifications
Material | Silica Gel |
Quantity | 500 g |
Mesh Size | 60 to 200 Mesh |
Packaging | Wide Mouth Amber Glass Bottle |
Grade | BAKER ANALYZED™ Reagent |
Chemical Name or Material | Silica Gel |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'