Learn More
Sartorius™ Silencer Unit
For use with Sartorius safety weighing cabinets.
Brand:  Sartorius™ YWCG03
Code : NEW
Additional Details : Weight : 1.00000kg
Product Code. 15341485
  • £499.00 / Each
In Stock
to see stock.
Description
Description
  • Attaches to fan filter/exhaust box
  • Accessory item
Specifications
Specifications
Safety Weighing Cabinets
To fan exhaust
Silencer Unit