Learn More
Ambion™ Silencer™ Select Pre-Designed siRNA
Silencer™ Select Pre-Designed siRNA
Brand: Ambion™ 4444246
Code : NEW
Additional Details : Weight : 0.00450kg
Description
The Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry. A choice of Silencer™ Select siRNAs are available—Pre-Designed or Validated. Silencer™ Select Pre-Designed siRNAs are guaranteed-to-silence based on their proven design while the Silencer™ Select Validated siRNAs are functionally tested to reduce target gene expression. Sets of Silencer™ Select siRNA Libraries are also available in tubes or plate for high throughput screening. The Silencer™ Select Positive and Negative Control RNAs should be included in every siRNA experiment.The benefits of using Silencer™ Select siRNAs include:
Note: Silencer™ Pre-Designed & Validated siRNAs are designed using our first-generation algorithm and DO NOT include any chemical modifications. They are recommended for completing in vitro RNAi studies that were started using Silencer™ siRNAs. For new studies we recommend Silencer Select siRNAs.
Approximate Turnaround Times*
Specifications
Room temperature | |
Silencer™ Select siRNA | |
1 μmol |
Silencer™ Select siRNA, 1μmol, HPLC Purity, The Silencer Select siRNAs are Classic 21-mers Which Incorporate the Latest Improvements in siRNA Design, Off-target Effect Prediction Algorithms and Chemistry | |
HPLC |
For Research Use Only. Not for use in diagnostic procedures.