missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Silencer™ Select Custom Designed siRNA

Silencer™ custom siRNAs (unmodified 21-bp duplexes) are cost-effective RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types.

Brand:  Ambion™ 4399669

Code : NEW

Additional Details : Weight : 0.00450kg

 View more versions of this product

Product Code. 15541054

  • £516.00 / 40nmol
Estimated Shipment: 03-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Cost-effective RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types
  • Use the GeneAssist™ Custom siRNA Builder to design and order a custom siRNA
Specifications

Specifications

Contains dried siRNA and Nuclease-free Water (1.75 mL), shipped at ambient temperature. Upon receipt, store siRNAs in a non-frost-free freezer at or below -20°C
Knocking down gene expression in order to study protein function in a wide range of cell types
Custom Designed siRNA
40 nmol
Upon receipt, store in a non-frost-free freezer at or below -20°C
GeneAssist - Custom Designed Silencer™ Select siRNA
Silencer™ Select
HPLC
Ambient
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.