missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Silberbrand™ Borosilicate Burettes with Straight Stopcock

Brand:  Brand™ 12013

Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Código de producto. 11804050

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Ver productos alternativos »


Especificaciones

Especificaciones

DIN EN ISO 385
470mm
Glass
Burette
0.05mL
Borosilicate
10mL
Videos