missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Silberbrand™ Borosilicate Burettes with Straight Stopcock

Brand:  Brand™ 12018

Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11824050

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

DIN EN ISO 385
790mm
Glass
Burette
0.1mL
Borosilicate
50mL
Videos