Learn More
BRAND™ SILBERBRAND Automatic Burette with PTFE Stockcock
SILBERBRAND Automatic Burette equipped with an i-stopcock and lateral stopcock.
Brand: BRAND™ 22332
Code : NEW
Additional Details : Weight : 1.34000kg
Description
- Error limits according to class B, DIN EN ISO 385
- Interchangeable keys
- Calibrated to deliver (TD, Ex)
- With glass-key in the titration and intermediate stopcock
- Automatic zeroing
- Intermediate stopcock to recirculate residual liquid
Specifications
25 mL | |
2000 mL bottle | |
12 months |
0.1 mL | |
i-stopcock, amber lateral stopcock, PTFE |