Learn More
Siguazodan, Tocris Bioscience™
PDE3 inhibitor
Brand:  Tocris (Bio-Techne) 114850
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13488733
  • £557.72 / 50mg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
Phosphodiesterase inhibitor, selective for PDE3 (IC50 = 117 nM).
Specifications
Chemical Identifiers
115344-47-3
284.323
N-Cyano-N'-methyl-N''-[4-(1,4,5,6-tetrahydro-4-methyl-6-oxo-3-pyridazinyl)phenyl]guanidine
C14H16N6O
NUHPODZZKHQQET-UHFFFAOYSA-N
72124
Specifications
Siguazodan
>99%
N-Cyano-N'-methyl-N''-[4-(1,4,5,6-tetrahydro-4-methyl-6-oxo-3-pyridazinyl)phenyl]guanidine
CC1CC(=O)NN=C1C2=CC=C(C=C2)NC(=NC)NC#N
284.323
284.32
115344-47-3
C14H16N6O
NUHPODZZKHQQET-UHFFFAOYSA-N
1-cyano-2-methyl-3-[4-(4-methyl-6-oxo-4,5-dihydro-1H-pyridazin-3-yl)phenyl]guanidine
72124