missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Siglec-F Antibody, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | Siglec-F |
---|---|
Clone | 238023 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Siglec-F Monoclonal specifically detects Siglec-F in Mouse samples. It is validated for Western Blot.Specifications
Siglec-F | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q920G3 | |
27181 | |
Mouse myeloma cell line NS0-derived recombinant mouse Siglec-F Asp18-Thr437 (predicted) Accession # Q920G3 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Siglec-F in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Siglec-5, -7, -9, -10, or recombinant mouse Siglec-2, or -3 is observed. |
238023 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
SAF2, sialic acid binding Ig like lectin 8, SIGLEC-8, SIGLEC8L, SiglecF | |
Siglecf | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only