missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Siglec-F Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | Siglec-F |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Siglec-F Polyclonal specifically detects Siglec-F in Mouse samples. It is validated for Western Blot.Specifications
Siglec-F | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
SAF2, sialic acid binding Ig like lectin 8, SIGLEC-8, SIGLEC8L, SiglecF | |
Siglecf | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q920G3 | |
27181 | |
Mouse myeloma cell line NS0-derived recombinant mouse Siglec-F Asp18-Thr437 Accession # Q920G3 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Siglec-F in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse (rm) Siglec-2, rmSiglec-3, recombinant human (rh) Siglec-5, rhSiglec-7, rhSiglec-9, and rhSiglec-10 is observed. |
For Research Use Only