Learn More
Fisherbrand™ S 50 R Sieve Holder
Supports analyzing sieves for the S 50 R Ultrasonic Cleaning Unit. Fisherbrand™ S 50 R Sieve Holders facilitate the special “sieve cleaning” program using two ultrasonic modes alternately, creating powerful cleaning pulses and perfect results.
Brand:  Fisherbrand™
Code : A0
Additional Details : Weight : 0.10000kg
Product Code. 12619355
  • £56.80 / Each
Estimated Shipment: 28-05-2024
to see stock.
Add to basket
Description
Description
  • Made of stainless steel
Specifications
Specifications
Ultrasonic Bath, S 50 R
Stainless Steel