Learn More
Kinematica™ Stainless Steel Sieve for Polymix™ MFC 90 D Mill
Depending on the sample, even finer results can be achieved by grinding with two or more sieves with progressively smaller hole sizes.
Brand:  Kinematica™ 35095020
Code : 04
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10617693
  • £154.00 / Each
Estimated Shipment: 16-07-2024
to see stock.
Add to basket
Specifications
Sieve
PX MFC 90 D
Stainless Steel
2.0 mm