Learn More
Tocris Bioscience™ SIB 1893
mGlu5 antagonist; also positive allosteric modulator at mGlu4 receptors
Brand:  Tocris Bioscience™ 1214/10
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13438903
  • £128.25 / 10mg
Estimated Shipment
08-07-2019
Description
Description
A highly selective non-competitive antagonist for the metabotropic glutamate mGlu5 receptor subtype; displays an IC50 value of 0.3 μM at hmGlu5, compared with > 100 μM at hmGlu1b, hmGlu2, hmGlu6, hmGlu7 and hmGlu8. Centrally active upon systemic administration in vivo. Positive allosteric modulator at mGlu4 receptors.
Specifications
Specifications
SIB 1893
SIB 1893
195.26