missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SHP-1 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | SHP-1 |
---|---|
Clone | 255402 |
Concentration | LYOPH |
Dilution | Immunocytochemistry 3-25 ug/mL |
Applications | Immunocytochemistry |
Description
SHP-1 Monoclonal specifically detects SHP-1 in Human samples. It is validated for Immunocytochemistry.Specifications
SHP-1 | |
LYOPH | |
Immunocytochemistry | |
Unconjugated | |
Rat | |
Human | |
P29350 | |
5777 | |
E. coli-derived recombinant human SHP-1 Ala205-Lys595 Accession # P29350 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human SHP-1. |
255402 | |
Immunocytochemistry 3-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HCP, HCPH, HPTP1C, PTP1C, PTP-1C, PTPN6, SHP1, SHP-1L, SH-PTP1 | |
PTPN6 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only