Learn More
SHOWA™ NSK24™ Blue Chemical Resistant Nitrile Coated Gloves
Catalog Number 12980795
Click to view available options
Size 11
Watertight, knit-lined nitrile glove provides protection from a wide array of solvents, oils and acids.
Protection from a wide array of solvents, oils and acid.
  • Flexibility and dexterity not found in a heavier- mil glove
  • Rough-finish texture delivers a sure grip in even the most oily and wet applications
  • Interlock liner ensures an added degree of comfort and liner protection
  • Color: Blue
Compliance
Data Testing is available upon request.