Learn More
SHOWA™ NSK24™ Gloves
A super flexible, watertight knit-lined nitrile glove
Brand:  SHOWA™ BSTNSK243
Code : 50
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 12970795
  • £640.00 / Pack of 72
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
Provides excellent protection from a wide array of solvents, oils and acids.
  • Offer flexibility and dexterity not found in a heavier-mil glove
  • Rough-finish texture delivers a sure grip in even the most oily and wet applications
  • Interlock liner ensures an added degree of comfort and liner protection
  • Color: Blue
Specifications
Specifications
Rough
Suitable for use in Oily greasy applications, refineries, sewer cleaning, deep sea fishing, wet cement handling, protection from hexavalent chromium, protection from fuels, painting, masonry and food handling applications
Nitrile
21 CFR 170-199, CE
35.5cm
Gauntlet
Cotton
10
Blue
Videos