missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ N-DEX™ 7700PF Nighthawk Gloves

Catalog Number p-2781791
Click to view available options
Size:
Small
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Black, powder free static-dissipative gloves are ideal for a wide variety of industrial applications.

  • ESD properties
  • 100% chemical-resistant nitrile
  • Accelerator and powder free
  • Textured fingertips for enhanced tactility
  • Made in the U.S.A.
  • Ideal for a wide variety of applications including laboratory, food processing/handling, automotive, law enforcement and electronic assembly
Certifications/Compliance FDA 21 CFR 170-199
Certifications/Compliance (Europe) Cat. III, CE Compliant 0123, EN 374-2, EN 374-3
Applications Industrial, Laboratory
AQL 1.5
Color Black
Disposable Single-use
Exterior Finish Partially Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Small
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
Model 7700PFTS
Length (English) 9.5 in.
Length (Metric) 24 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'