missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ N-DEX™ 9500PF Fluorescent Green Nitrile Gloves

Catalog Number 12960615
Click to view available options
Size:
Small
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Fluorescent green, powder free nitrile disposable gloves are 11 inches in length and provide high resistance to puncture, tearing, abrasion, and chemical splash.

  • High visibility fluorescent green color
  • Free from common accelerators found in nitrile gloves
  • Thicker barrier protection, 5 mil
  • Second skin fit and feel
  • Rolled cuff
  • Smooth finish for tactile sensitivity
  • Powder free
Certifications/Compliance ASTM F 1671, NFPA 1999, 21 CFR 170-199
Chemotherapeutic Rated No
Applications Industrial, Laboratory
AQL 1.5
Color Hi-vis Green
Disposable Single-use
Exterior Finish No Texture
Finger Thickness (English) 5 mil
Finger Thickness (Metric) 0.12 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 5 mil
Palm Thickness (Metric) 0.12 mm
Powdered Powder Free
Size Small
Thickness (English) 5 mil
Thickness (Metric) 0.12 mm
FDA 510 (k) Yes
Model 9500PFSM
Series N-DEX™ 9500PF
Length (English) 11 in.
Length (Metric) 28 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'