missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ Atlas™ 460 Double-Dipped Gloves

Brand:  SHOWA™ SHO4604

Code : S4

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12950145

  • £22.90 / Each


Estimated Shipment
01-03-2018
Add to basket

Specifications

Specifications

Full
Launder at 40°C using a neutral Detergent
Gauntlet
PVC (Polyvinyl Chloride)
10
30.5cm
Level 2
Rough
manufacturing, commercial fishing, cold storage, construction, de-icing, fuel transport, agriculture
Acrylic
Orange
Videos