Learn More
Sartorius™ Shock Absorbers for Analytical Balance
Pinwheel-shaped footpads for your analytical balance. Sartorius™ Shock Absorbers for Analytical Balance reduce vibrations that can disrupt balance operations.
Brand:  Sartorius™ 69B20006
Code : NEW
Additional Details : Weight : 0.99000kg
Product Code. 10735889
  • £22.05 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Attach to the bottom of your analytical balance
  • Reduce vibrations
  • Pinwheel-shaped
Specifications
Specifications
Shock absorbers
Analytical balance
Videos