missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ Sheet-Steel Shelf

Brand:  Asecos™ 30059

Code : NEW

Additional Details : Weight : 2.00000kg

 View more versions of this product

Product Code. 15377831

  • £41.82 / Each


Estimated Shipment
03-08-2018
Add to basket

Specifications

Specifications

For RAL 7035
Shelf
740mm
30mm
75kg
Steel
Videos