Learn More
SHC3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561274
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: HPYYNSIPSK MPPPGGFLDT RLKPRPHAPD TAQFAGKEQT YYQGRHLGDT FGEDWQQTPL RQGSSDIYST PEGKLHVAPT GEAPTYVNTQ QIPP Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.
SHC3 Polyclonal antibody specifically detects SHC3 in Human samples. It is validated for Immunohistochemistry.Specifications
SHC3 | |
Polyclonal | |
Unconjugated | |
SHC3 | |
neuronal Shc; Nshc; N-Shc; Protein Rai; RAI; SH2 domain protein C3; SHC (Src homology 2 domain containing) transforming protein 3; SHC adaptor protein 3; SHC3; Shc3 p51; SHCC; SHC-transforming protein 3; SHC-transforming protein C; src homology 2 domain-containing transforming protein C3; src homology 2 domain-containing-transforming protein C3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
53358 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q92529 | |
SHC3 | |
Recombinant protein corresponding to Human SHC3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only