Learn More
Sharpsguard™ anatomical 22 Waste Container
For the safe disposal of anatomical and placenta waste. Sharpsguard™ anatomical 22 Waste Containers are made from rigid polypropylene with a strong wall thickness.
Brand: Daniels Healthcare DD320
Code : 02
Additional Details : Weight : 0.80000kg
Description
- Offers outstanding protection
- When container is full to the fill line it must be locked and disposed of safely
- All have clear British Standard markings, user instructions and a visible fill line
Specifications
Anatomical Waste | |
21.01 L | |
Red, Yellow |