Learn More
Sharpsguard™ anatomical 11.5 Waste Container
For the safe disposal of anatomical and placenta waste. Sharpsguard™ anatomical 11.5 Waste Containers each come with 5 placenta bags.
Brand: Daniels Healthcare DD910
Code : 02
Additional Details : Weight : 0.90000kg
Description
- Rigid polypropylene construction and wall thickness
- Offers outstanding protection
- When container is full to the fill line it must be locked and disposed of safely
- All have clear British Standard markings, user instructions and a visible fill line
Specifications
Anatomical Waste | |
10.91 L | |
Red, Yellow |