missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Single Cytofunnel™ with Brown Filter Cards
Hold sample volumes of 0.4mL or less with the Epredia™ Single Cytofunnel™ with Brown Filter Cards.
Brand: Epredia™ 5991043
Code : 50
Additional Details : Weight : 0.45360kg
Description
- Includes disposable sample chamber with brown pre-attached filter card and cap
- Shandon™ Single Cytofunnel™ with Brown Filter Cards requires Epredia™ Cytoclips™
- Provides a cell deposition area of 6 mm (28 mm squared)
Specifications
50/Box | |
Single-use | |
Cytofunnel with Brown Filter Card | |
0.4 mL |