missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Shandon™ Acid-Resistant Biopsy Bags

Reduce risk of small specimen loss with Thermo Scientific™ Shandon™ Acid-Resistant Biopsy Bags.

Brand:  Thermo Scientific™ 6774051

Code : 50

Additional Details : Weight : 0.45360kg

 View more versions of this product

Product Code. 12677946

  • £144.44 / Pack of 250


Estimated Shipment
09-02-2018
Add to basket

Description

Description

  • Allow unrestricted fluid movement around tissue
  • Reduce reagent carryover and pressure artifact often found with sponges
  • Remove specimen rapidly with convenient peel-open bag

Dimensions:

  • Small Biopsy Bags measure 1.2 × 1.7″ (31 × 43mm)
  • Medium Biopsy Bags measure 1.8 × 2.6″ (45 × 65mm)
  • Large Biopsy Bags measure 3 × 3.5″ (75 × 85mm)
Specifications

Specifications

White
31mm
Nylon|Acid-Resistant
Biopsy Bag
43mm
Small
Videos
Documentation

Technical Documents