missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Self-Adhesive Graduation
Click to view available options
For Use With: 30L wide necked jerrycans
Description
- White and blue graduations
- Self-adhesive
- For use with wide-necked jerrycans
Specifications
Product Type | Graduation |
Description | Self adhesive graduation marks for jerrycans 30 L |
For Use With (Equipment) | 30 L Jerricans |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'